Learn More
Cayman Chemical GGT Substrate 1ea
SDP
Catalog No. 502978358
Encompass_Preferred
GGT substrate has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).
Catalog No. 50-297-8358 Supplier Cayman Chemical Supplier No. 4005161 EA
GGT substrate has been tested and formulated to work exclusively with Cayman’s γ-Glutamyl Transferase Activity Assay Kit (Item No. 702210).