Learn More
Santa Cruz Biotechnology GGT1 Antibody (F-7) Alexa Fluor® 647, 200 µg/ml
Catalog No. NC3961003
Encompass
GGT1 Antibody (F-7) is a mouse monoclonal IgG2b κ GGT1 antibody provided at 200 µg/mlspecific for an epitope mapping between amino acids 351-389 within an internal region of GGT1 of mouse originGGT1 Antibody (F-7) is recommended for detection of GGT1 heavy chain of mouse and rat origin by WB, IP, IF, IHC(P) and ELISAAnti-GGT1 Antibody (F-7) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM
Catalog No. NC3961003 Supplier Santa Cruz Biotechnology Supplier No. SC374495 AF647
GGT1 Antibody (F-7) is a mouse monoclonal IgG2b κ GGT1 antibody provided at 200 µg/mlspecific for an epitope mapping between amino acids 351-389 within an internal region of GGT1 of mouse originGGT1 Antibody (F-7) is recommended for detection of GGT1 heavy chain of mouse and rat origin by WB, IP, IF, IHC(P) and ELISAAnti-GGT1 Antibody (F-7) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM