Learn More
Aviva Systems Biology Corp GGT2 Antibody - N-terminal region (ARP49227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GGT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GGT2. Alternate names: GGT, GGT 2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-407