Learn More
Medchemexpress LLC GGTI-2154 (hydrochloride) | 478908-50-8 | 98.1% | 456.97 | 25 MG
SDP
Catalog No. 5000393963
Encompass_Preferred
GGTI-2154 hydrochloride is a potent and selective inhibitor of geranylgeranyltransferase I (GGTase I), with an IC50 of 21 nM. It demonstrates over 200-fold selectivity for GGTase I compared to FTase (IC50=5600 nM). This product is suitable for cancer research. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of geranylgeranyltransferase I (GGTase I) with an IC50 of 21 nM.
  • Shows more than 200-fold selectivity for GGTase I over FTase (IC50=5600 nM).
  • Can be used for cancer research.
  • Induces breast tumor regression in MMTV-ν-Ha-Ras transgenic mice (100 mg/kg/day; s.c. for 14 days).
  • Inhibits A-549 tumor growth in nude mice by 60% (50 mg/kg/day; i.p. for 50 days).
Catalog No. 50-003-93963 Supplier Medchemexpress LLC Supplier No. 25MGHY16229A
GGTI-2154 hydrochloride is a potent and selective inhibitor of geranylgeranyltransferase I (GGTase I), with an IC50 of 21 nM. It demonstrates over 200-fold selectivity for GGTase I compared to FTase (IC50=5600 nM). This product is suitable for cancer research. It is for research use only and not sold to patients.
  • Potent and selective inhibitor of geranylgeranyltransferase I (GGTase I) with an IC50 of 21 nM.
  • Shows more than 200-fold selectivity for GGTase I over FTase (IC50=5600 nM).
  • Can be used for cancer research.
  • Induces breast tumor regression in MMTV-ν-Ha-Ras transgenic mice (100 mg/kg/day; s.c. for 14 days).
  • Inhibits A-549 tumor growth in nude mice by 60% (50 mg/kg/day; i.p. for 50 days).