Learn More
Selleck Chemical LLC GGTI 298 TFA salt 1mg 1217457-86-7
SDP
Supplier:  Selleck Chemical LLC S74661MG
Encompass_Preferred
GGTI 298 is a geranylgeranyltransferase I inhibitor with ability to arrest human tumor cells in the G1 phase of the cell cycle and induce apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4820