Learn More
Selleck Chemical LLC GGTI 298 TFA salt
SDP
Catalog No. 5000001370
Encompass_Preferred
GGTI 298 is a geranylgeranyltransferase I inhibitor with ability to arrest human tumor cells in the G1 phase of the cell cycle and induce apoptosis
Catalog No. 50-000-01370 Supplier Selleck Chemical LLC Supplier No. S746625MG
Add to Cart
Add to Cart
GGTI 298 is a geranylgeranyltransferase I inhibitor with ability to arrest human tumor cells in the G1 phase of the cell cycle and induce apoptosis