Learn More
Selleck Chemical LLC GGTI 298 TFA salt
SDP
Supplier:  Selleck Chemical LLC S7466100MG
Encompass_Preferred
GGTI 298 is a geranylgeranyltransferase I inhibitor with ability to arrest human tumor cells in the G1 phase of the cell cycle and induce apoptosis
Catalog No. 50-000-04436