Learn More
Cayman Chemical GGTI 298 trifluoroacetate sa
SDP
Catalog No. 502969623
Encompass_Preferred
A CAAX peptidomimetic that selectively inhibits geranylgeranyltransferase I; arrests human tumor cells (IC50 = 10 µM for A549 cells) in G0/G1 and induces apoptosis by inhibiting proteasome activity and up-regulating the expression of the cyclin-dependent kinase inhibitor p21
Catalog No. 50-296-9623 Supplier Cayman Chemical Supplier No. 16176500 UG
A CAAX peptidomimetic that selectively inhibits geranylgeranyltransferase I; arrests human tumor cells (IC50 = 10 µM for A549 cells) in G0/G1 and induces apoptosis by inhibiting proteasome activity and up-regulating the expression of the cyclin-dependent kinase inhibitor p21