Learn More
Medchemexpress LLC HY-15871 1mg Medchemexpress, GGTI298 (Trifluoroacetate) CAS:1217457-86-7 Purity:>98%
SDP
Catalog No. 502081810
Encompass_Preferred
Medchemexpress, HY-15871 1mg GGTI298 (Trifluoroacetate) CAS:1217457-86-7 GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM; little effect on Ha-Ras with IC50 of >20 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1810 Supplier Medchemexpress LLC Supplier No. HY158711MG
Medchemexpress, HY-15871 1mg GGTI298 (Trifluoroacetate) CAS:1217457-86-7 GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor, which can inhibit Rap1A with IC50 of 3 μM; little effect on Ha-Ras with IC50 of >20 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.