Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GGTI298 Trifluoroacetate | 1217457-86-7 | 99.9% | 593.66 | 5 MG
SDP

Catalog No. 5000364367
Encompass_Preferred

GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor. It can inhibit Rap1A with an IC50 of 3 μM, while showing little effect on Ha-Ras with an IC50 of >20 μM. This compound appears as a white to yellow solid.

  • Significantly reduces cAMP agonist-stimulated apical K+ conductance.
  • Abolishes NF-κB activation, leading to sensitization of DR5-dependent apoptosis.
  • Activates NF-κB and inhibits Akt.
  • Reduces fluid accumulation in mouse ileal loop experiments.
  • Store at 4°C, sealed, away from moisture.
  • Soluble in DMSO at 100 mg/mL.

Catalog No. 50-003-64367 Supplier Medchemexpress LLC Supplier No. 5MGHY15871
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GGTI298 Trifluoroacetate is a CAAZ peptidomimetic geranylgeranyltransferase I (GGTase I) inhibitor. It can inhibit Rap1A with an IC50 of 3 μM, while showing little effect on Ha-Ras with an IC50 of >20 μM. This compound appears as a white to yellow solid.

  • Significantly reduces cAMP agonist-stimulated apical K+ conductance.
  • Abolishes NF-κB activation, leading to sensitization of DR5-dependent apoptosis.
  • Activates NF-κB and inhibits Akt.
  • Reduces fluid accumulation in mouse ileal loop experiments.
  • Store at 4°C, sealed, away from moisture.
  • Soluble in DMSO at 100 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.