Learn More
Aviva Systems Biology Corp GGTLA4 Antibody - C-terminal region (ARP41639_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41639_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GGTLA4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human GGTLA4. Alternate names: RP5-831C21.2, MGC50550, dJ831C21.2, GGTL6, GGTLA3, GGTLA4, dJ831C21.1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57929