Learn More
Medchemexpress LLC HY-P7183 50ug Medchemexpress, GH, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP718350UG
Encompass_Preferred
Medchemexpress, HY-P7183 50ug GH, Human CAS: Recombinant Human Growth Hormone is a peptide hormone that stimulates growth, cell reproduction, and cell regeneration, which can be used for the treatment of growth disorders in children. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4359