Learn More
Genscript Corporation GH HumanCHO-expressed
Supplier:  Genscript Corporation Z0300510
Encompass
Growth Hormone (GH) is a member of the somatotropin/prolactin family which play an important role in growth control The human GH cDNA encodes a 217 amino acid (aa) and the first 26 aa is a signal peptide By alternative splicing at least four isoforms of GH have been identified The major role of GH in stimulating body growth is to stimulate the liver and other tissues to secrete IGF-1 GH stimulates both the differentiation and proliferation of myoblasts and also stimulates amino acid uptake and protein synthesis in muscle and other tissues
Catalog No. 50-000-58808