Learn More
Eurofins Discoverx Growth Hormone Releasing Factor, 0.5 mg. (92-1058)
Supplier:  Eurofins Discoverx 921058
Encompass
Growth Hormone Releasing Factor (GRF or GHRF) is a preproprotein produced in the hypothalamus that acts on the adenohypophyse to stimulate growth hormone secretion from the pituitary gland. This activity may stimulate tumor growth. GRF levels are regulated by somatostatin, growth hormone, and insulin-like growth factor 1. GRF acts as an agonist for GHRHR. Provided as 0.5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9599