Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2688MSM3P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: GH/1450. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b. Positive Control: Pituitary cells or Pituitary. Cellular Staining: Cytoplasmic. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: A recombinant human Growth Hormone (GH) fragment (aa58-187). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 20kDa.Chromosome: 17q24.2. Gene Symbol: GH1. Gene ID: 2688. Ensemble: ENSG00000259384. Uniprot: NP_072053.
Catalog No. 50-188-4733