Learn More
Aviva Systems Biology Corp GH1 Antibody - C-terminal region (ARP41763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GH1. It was validated on Western Blot. . Alternate names: GH, GH-N, GHN, IGHD1B, hGH-NApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-284-953