Learn More
Aviva Systems Biology Corp GH2 Antibody - middle region (ARP42013_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42013_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GH2. Alternate names: GH-V, GHL, GHV, hGH-VApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58080