Learn More
ACROBiosystems Varicella zoster virus (strain Oka vaccine) gH&gL Protein, Twin-Strep Tag&His Tag (MALS verified)
Catalog No. 502719592
Encompass_Preferred
The heterodimer glycoprotein H-glycoprotein L is required for the fusion of viral and plasma membranes leading to virus entry into the host cell. Acts as a functional inhibitor of gH and maintains gH in an inhibited form. Upon binding to host integrins, gL dissociates from gH leading to activation of the viral fusion glycoproteins gB and gH.
Catalog No. 50-271-9592 Supplier ACROBiosystems Supplier No. GHLV5283500UG
The heterodimer glycoprotein H-glycoprotein L is required for the fusion of viral and plasma membranes leading to virus entry into the host cell. Acts as a functional inhibitor of gH and maintains gH in an inhibited form. Upon binding to host integrins, gL dissociates from gH leading to activation of the viral fusion glycoproteins gB and gH.