Learn More
Aviva Systems Biology Corp Ghitm Antibody - middle region (ARP47019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ghitm. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1010001P14Rik, C77840, PTD010, MICS1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-280