Learn More
Cayman Chemical GHK-Cuacetate 100mg
SDP
Catalog No. 502940327
Encompass_Preferred
A complex of the tripeptide Gly-His-Lys and a copper(II) ion that has wound healing and anti-inflammatory activities; increases proliferation and the levels of collagen and secreted pro-MMP-2 in isolated human fibroblasts at 1 nM; increases levels of collagen, GAGs, and the expression of decorin in the wound tissue of rats at 2 mg; decreases LPS-induced increases in levels of ROS, IL-6, and TNF-α in RAW 264.7 cells at 10 µM; prevents LPS-induced decreases in lung SOD activity and GSH levels and reduces LPS-induced increases in the number of cells and the level of total protein in BALF in a mouse model of acute lung injury
Catalog No. 50-294-0327 Supplier Cayman Chemical Supplier No. 28259100 MG
A complex of the tripeptide Gly-His-Lys and a copper(II) ion that has wound healing and anti-inflammatory activities; increases proliferation and the levels of collagen and secreted pro-MMP-2 in isolated human fibroblasts at 1 nM; increases levels of collagen, GAGs, and the expression of decorin in the wound tissue of rats at 2 mg; decreases LPS-induced increases in levels of ROS, IL-6, and TNF-α in RAW 264.7 cells at 10 µM; prevents LPS-induced decreases in lung SOD activity and GSH levels and reduces LPS-induced increases in the number of cells and the level of total protein in BALF in a mouse model of acute lung injury