Learn More
Rockland Immunochemicals Ghost Red Blood Cells, Non-Sterile, 99% Hemoglobin Removal, 5 mL
SDP
Catalog No. NC3663713
Encompass_Preferred
Ghost Red Blood Cells are a unique model system derived from goat blood, ideal for studying biological membranes and phospholipid metabolism. This product aids in advancing drug delivery technologies, such as nanoerythrocytes. Prepared through a meticulous washing process, it retains erythrocyte membrane integrity while eliminating 99% of hemoglobin. Available in a 5 mL liquid form, it is non-sterile and requires storage at 4°C prior to opening.
Catalog No. NC3663713 Supplier Rockland Immunochemicals Supplier No. R7040055ML
Ghost Red Blood Cells are a unique model system derived from goat blood, ideal for studying biological membranes and phospholipid metabolism. This product aids in advancing drug delivery technologies, such as nanoerythrocytes. Prepared through a meticulous washing process, it retains erythrocyte membrane integrity while eliminating 99% of hemoglobin. Available in a 5 mL liquid form, it is non-sterile and requires storage at 4°C prior to opening.