Learn More
MSC GHP GROUP Forced Air Heater, 135,000 BTU, Multi-Fuel, With Thermostat
Catalog No. 5000509177
Encompass
This forced air heater provides powerful and efficient heating for areas up to 3,200 square feet. It operates on various fuels including diesel, kerosene, JP-8, and Jet A, offering flexibility in fuel sourcing. Equipped with a built-in thermostat for precise temperature control and electronic spark ignition, it ensures reliable starting. The robust steel housing provides durability, and the unit is UL and cCSAus certified, ensuring safety and quality standards are met.
  • Heats areas up to 3,200 square feet.
  • Compatible with multiple fuel types: diesel, kerosene, JP-8, and Jet A.
  • Features a built-in thermostat for accurate temperature management.
  • Electronic spark ignition for dependable starting.
  • Durable steel housing.
  • 10 gallon fuel capacity provides 10 hours of operating time.
  • 500 CFM airflow.
  • UL and cCSAus certified.
Catalog No. 50-005-09177 Supplier MSC Supplier No. 99892564
This forced air heater provides powerful and efficient heating for areas up to 3,200 square feet. It operates on various fuels including diesel, kerosene, JP-8, and Jet A, offering flexibility in fuel sourcing. Equipped with a built-in thermostat for precise temperature control and electronic spark ignition, it ensures reliable starting. The robust steel housing provides durability, and the unit is UL and cCSAus certified, ensuring safety and quality standards are met.
  • Heats areas up to 3,200 square feet.
  • Compatible with multiple fuel types: diesel, kerosene, JP-8, and Jet A.
  • Features a built-in thermostat for accurate temperature management.
  • Electronic spark ignition for dependable starting.
  • Durable steel housing.
  • 10 gallon fuel capacity provides 10 hours of operating time.
  • 500 CFM airflow.
  • UL and cCSAus certified.