Learn More
Biomatik Corporation Ghrelin (Ghrl) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29823
Encompass_Preferred
Ghrelin (Ghrl) Monoclonal Antibody is a Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Dog (Canis familiaris; Canine) Monoclonal Antibody to Ghrelin (GHRL). The Ghrelin (Ghrl) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Dog (Canis familiaris; Canine). Applications: WB, IHC, ICC, IP. Target Synonyms: Ghrelin (GHRL). Accession Number: Q9UBU3. Gene ID: 51738. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5668