Learn More
CPC Scientific H-Gly-Ser-Ser(n-octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GHRE001A
Encompass
Growth-hormone releasing acylated peptide.SEQUENCE: H-Gly-Ser-Ser(n-octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GSS(n-octanoyl)-FLSPEHQRVQQRKESKKPPAKLQPRMOLECULAR FORMULA: C149H249N47O42MOLECULAR WEIGHT: 3370.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [258279-04-8]SYNONYMS: RESEARCH AREA: GastrointestinalREFERENCES:M. Kojima, et al., Nature, 402, 656 (1999)SKU(s): GHRE-001A, GHRE-001B
Catalog No. 50-210-9323