Learn More
Carbosynth LLC. Ghrelin (Human), 1mg, MW: 3370.94, Molecular formula: C149H249N47O42, CAS# 258279-04-8
SDP
Supplier:  Carbosynth LLC. PGH3741PI1MG
Encompass_Preferred
Synonyms: H-Gly-Ser-Ser(n-Octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH, Application: Orexigenc and GH Releasing Peptide, Salt Form:, Trifluoroacetate, Storage Temperature: -20 °C
Catalog No. 50-168-6519