Learn More
CPC Scientific H-Gly-Ser-Ser-Phe-Leu-Ser-Pro-Glu-His-Gln-Arg-Val-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GHRE005B
Encompass
Small synthetic molecules, called growth-hormone secretagogues (GHSs), that stimulate the release of growth hormone (GH) from the pituitary through a G protein-coupled receptor (GHS-R). It has been demonstrated, however, to not possess the pituitaric and pancreatic endocrine activities of octanoylated ghrelin in humans.ONE-LETTER SEQUENCE: GSSFLSPEHQRVQQRKESKKPPAKLQPRMOLECULAR FORMULA: C141H235N47O41MOLECULAR WEIGHT:3244.71STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [313951-59-6], RESEARCH AREA: GastrointestinalREFERENCES: Family Literatures:M.Kojima et al., Nature, 402, 656(1999) ; C. Dieguez and F.F.Casanueva,Eur.J. Endocrinol., 142, 413(2000)
Catalog No. 50-210-9332