Learn More
CPC Scientific H-Gly-Ser-[Ser(n-octanoyl)]-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GHRE003A
Encompass
Endogenous growth-hormone releasing acylated peptide with novel regulatory mechanisms.SEQUENCE: H-Gly-Ser-[Ser(n-octanoyl)]-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: GS-[S(n-octanoyl)]-FLSPEHQKAQQRKESKKPPAKLQPRMOLECULAR FORMULA: C147H245N45O42MOLECULAR WEIGHT: 3314.8STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [258338-12-4]SYNONYMS: RESEARCH AREA: GastrointestinalREFERENCES:M. Kojima, et al., Nature, 402, 656 (1999)SKU(s): GHRE-003A, GHRE-003B
Catalog No. 50-210-9327