Learn More
CPC Scientific H-Gly-Ser-Ser(palmitoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific GHRE004B
Encompass
ONE-LETTER SEQUENCE: GSS(palmitoyl)FLSPEHQKAQQRKESKKPPAKLQPR MOLECULAR FORMULA: C155H261N45O42MOLECULAR WEIGHT:3427.1STORAGE CONDITIONS: -20 5CSYNONYMS: hunger hormone, lenomorelinRESEARCH AREA: ObesityREFERENCES: M. Kojima, et al., Nature, 402, 656 (1999)
Catalog No. 50-210-9330