Learn More
Carbosynth LLC. Ghrelin (Rat) (0.1mg vial), 1ea, MW: 3314.8, Molecular formula: C147H245N45O42, CAS# 258338-12-4
SDP
Catalog No. 501686538
Encompass_Preferred
Synonyms: Gly-Ser-Ser(n-Octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: Appetite Stimulating Peptide with Energy Homeostasis Regulation M. Kojima, H. Hosoda, Y. Date, M. Nakazato, H. Matsuo, and K. Kangawa, Nature, 402, 656 (1999). (Original), C. Dieguez and F.F. Casanueva, Eur. J. Endocrinol., 142, 413 (2000). (Review), G. Muccioli, M. Tschöp, M. Papotti, R. Deghenghi, M. Heiman, and E. Ghigo, Eur. J. Pharmacol., 440, 235 (2002). (Review)
Catalog No. 50-168-6538 Supplier Carbosynth LLC. Supplier No. PGH4373S0.1MG
Synonyms: Gly-Ser-Ser(n-Octanoyl)-Phe-Leu-Ser-Pro-Glu-His-Gln-Lys-Ala-Gln-Gln-Arg-Lys-Glu-Ser-Lys-Lys-Pro-Pro-Ala-Lys-Leu-Gln-Pro-Arg, Salt Form:, Trifluoroacetate Salt, Storage Temperature: -20°C, References: Appetite Stimulating Peptide with Energy Homeostasis Regulation M. Kojima, H. Hosoda, Y. Date, M. Nakazato, H. Matsuo, and K. Kangawa, Nature, 402, 656 (1999). (Original), C. Dieguez and F.F. Casanueva, Eur. J. Endocrinol., 142, 413 (2000). (Review), G. Muccioli, M. Tschöp, M. Papotti, R. Deghenghi, M. Heiman, and E. Ghigo, Eur. J. Pharmacol., 440, 235 (2002). (Review)