Learn More
CPC Scientific H-Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2 (acetate salt) 1MG
SDP
Supplier:  CPC Scientific GHRF001B
Encompass
Shortest GRF fragment with full biological activity.ONE-LETTER SEQUENCE: YADAIFTNSYRKVLGQLSARKLLQDIMSR-NH2MOLECULAR FORMULA: C149H246N44O42S1MOLECULAR WEIGHT:3358STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86168-78-7], RESEARCH AREA: HormonalREFERENCES: V.A. Lance et al., Biochem. Biophys. Res. Commun., 119, 265 (1984)
Catalog No. 50-210-9334