Learn More
CPC Scientific H-His-Ala-Asp-Ala-Ile-Phe-Thr-Ser-Ser-Tyr-Arg-Arg-Ile-Leu-Gly-Gln-Leu-Tyr-Ala-Arg-Lys-Leu-Leu-His-Glu-Ile-Met-Asn-Arg-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GHRF007A
Encompass
ONE-LETTER SEQUENCE: HADAIFTSSYRRILGQLYARKLLHEIMNR-NH2MOLECULAR FORMULA: C155H251N49O40S1MOLECULAR WEIGHT:3473.02STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [91826-20-9], SYNONYMS: Somatoliberin(1-29)amide[rat], somatoliberin fragment 1-29 amide, ratRESEARCH AREA: Hormonal
Catalog No. 50-210-9343