Learn More
Medchemexpress LLC Ghrf, mouse (TFA) | 125199-49-7 | 98.9% | 5032.74 | 5 MG
SDP
Catalog No. 5000399350
Encompass_Preferred
GHRF, mouse (TFA) is a 44-amino acid peptide that stimulates the release and synthesis of growth hormone. This product is for research use only and not intended for patient use.
  • 44-amino acid peptide
  • Stimulates growth hormone release and synthesis
  • High purity of 98.9%
  • Molecular weight of 5032.74
  • Increases growth hormone levels in LI-IGF-I-/- mice
Catalog No. 50-003-99350 Supplier Medchemexpress LLC Supplier No. HYP3596A5MG
GHRF, mouse (TFA) is a 44-amino acid peptide that stimulates the release and synthesis of growth hormone. This product is for research use only and not intended for patient use.
  • 44-amino acid peptide
  • Stimulates growth hormone release and synthesis
  • High purity of 98.9%
  • Molecular weight of 5032.74
  • Increases growth hormone levels in LI-IGF-I-/- mice