Learn More
CPC Scientific H-His-Ala-Asp-Ala-Ile-Phe-Thr-Ser-Ser-Tyr-Arg-Arg-Ile-Leu-Gly-Gln-Leu-Tyr-Ala-Arg-Lys-Leu-Leu-His-Glu-Ile-Met-Asn-Arg-Gln-Gln-Gly-Glu-Arg-Asn-Gln-Glu-Gln-Arg-Ser-Arg-Phe-Asn-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502109342
Encompass
ONE-LETTER SEQUENCE: HADAIFTSSYRRILGQLYARKLLHEIMNRQQGERNQEQRSRFNMOLECULAR FORMULA: C225H361N77O66S1MOLECULAR WEIGHT:5232.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86472-71-1], RESEARCH AREA: HormonalREFERENCES: J. Spiess et al., Nature, 303, 532 (1983)
Catalog No. 50-210-9342 Supplier CPC Scientific Supplier No. GHRF006B
ONE-LETTER SEQUENCE: HADAIFTSSYRRILGQLYARKLLHEIMNRQQGERNQEQRSRFNMOLECULAR FORMULA: C225H361N77O66S1MOLECULAR WEIGHT:5232.9STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86472-71-1], RESEARCH AREA: HormonalREFERENCES: J. Spiess et al., Nature, 303, 532 (1983)