Learn More
Aviva Systems Biology Corp GHRHR Antibody - C-terminal region (ARP41859_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41859_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GHRHR. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GHRHR. Alternate names: GHRFR, GHRHRpsv, GRFR, IGHD1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-58019