Learn More
CPC Scientific H-DAla-2-DNal-Ala-Trp-DPhe-Lys-NH2 0.5MG
SDP
Catalog No. 502109347
Encompass
Newly developed GHRP that has been studied in rats and produces a large plasma GH response in the presence of exogenous GH-releasing factor. This response was significantly augmented when somatostatin-antiserum was present. This peptide could be an alternative therapeutic agent in GH therapy.SEQUENCE: H-DAla-2-DNal-Ala-Trp-DPhe-Lys-NH2ONE-LETTER SEQUENCE: DAla-2-DNal-AW-DPhe-K-NH2MOLECULAR FORMULA: C45H55N9O6MOLECULAR WEIGHT: 818STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [158861-67-7]SYNONYMS: KP-102, Growth hormone-releasing peptide 2, Pralmorelin, (D-Ala1,D-2-Nal2)-GHRP-6RESEARCH AREA: HormonalREFERENCES:H.Sawada et al., Regul. Peptides, 53, 195 (1994)SKU(s): GHRF-009A, GHRF-009B
Catalog No. 50-210-9347 Supplier CPC Scientific Supplier No. GHRF009A
Newly developed GHRP that has been studied in rats and produces a large plasma GH response in the presence of exogenous GH-releasing factor. This response was significantly augmented when somatostatin-antiserum was present. This peptide could be an alternative therapeutic agent in GH therapy.SEQUENCE: H-DAla-2-DNal-Ala-Trp-DPhe-Lys-NH2ONE-LETTER SEQUENCE: DAla-2-DNal-AW-DPhe-K-NH2MOLECULAR FORMULA: C45H55N9O6MOLECULAR WEIGHT: 818STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [158861-67-7]SYNONYMS: KP-102, Growth hormone-releasing peptide 2, Pralmorelin, (D-Ala1,D-2-Nal2)-GHRP-6RESEARCH AREA: HormonalREFERENCES:H.Sawada et al., Regul. Peptides, 53, 195 (1994)SKU(s): GHRF-009A, GHRF-009B