Learn More
Cayman Chemical GHRP-6acetate 100mg
SDP
Catalog No. 502938886
Encompass_Preferred
A synthetic GH secretagogue and an agonist of GHS-R; inhibits binding of the GHS-R agonist MK-0677/ibutamoren (Ki = 1.9 nM) and ghrelin (Kd = 260 nM) to GSH-R type Ia and GSH-R, respectively; stimulates intracellular calcium mobilization (EC50 = 4.5 nM) and inositol phosphate production (EC50 = 0.83 nM) in cells expressing the human receptor; negative allosteric modulator of ghrelin signaling; induces release of GH, but not TSH, LH, or FSH, in isolated rat pituitary gland at 0.03 μg/ml; increases levels of GH, but not TSH, LH, FSH, or prolactin, in rat blood at 50 μg; increases food intake in rats at 0.3, 1, and 3 nmol
Catalog No. 50-293-8886 Supplier Cayman Chemical Supplier No. 27262100 MG
A synthetic GH secretagogue and an agonist of GHS-R; inhibits binding of the GHS-R agonist MK-0677/ibutamoren (Ki = 1.9 nM) and ghrelin (Kd = 260 nM) to GSH-R type Ia and GSH-R, respectively; stimulates intracellular calcium mobilization (EC50 = 4.5 nM) and inositol phosphate production (EC50 = 0.83 nM) in cells expressing the human receptor; negative allosteric modulator of ghrelin signaling; induces release of GH, but not TSH, LH, or FSH, in isolated rat pituitary gland at 0.03 μg/ml; increases levels of GH, but not TSH, LH, FSH, or prolactin, in rat blood at 50 μg; increases food intake in rats at 0.3, 1, and 3 nmol