Learn More
Uline GHS Pictogram Labels - Health Hazard, 1 x 1"", 500/roll
Catalog No. NC1376663
Encompass_Preferred
Label hazardous products to meet OSHA's Globally Harmonized System (GHS) requirements. Identify substances that may cause cancer, fertility issues or organ damage. Semi-gloss paper with permanent adhesive. Labels can be combined for quantity pricing.
Catalog No. NC1376663 Supplier Uline Supplier No. S21337
Label hazardous products to meet OSHA's Globally Harmonized System (GHS) requirements. Identify substances that may cause cancer, fertility issues or organ damage. Semi-gloss paper with permanent adhesive. Labels can be combined for quantity pricing.