Learn More
Enzo Life Sciences Ghrelin receptor GHS-R polyclonal antibody (100µg)
SDP
Catalog No. 502007059
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat Ghrelin Receptor (GHS-R). UniProt ID: O08725. Species reactivity: Rat. Creossreactivity: Predicted species reactivity based on epitope sequence homology: human, mouse, marmoset, rhesus macaque, platypus, horse. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.
Catalog No. 50-200-7059 Supplier Enzo Life Sciences Supplier No. ADI905792100
Add to Cart
Add to Cart
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat Ghrelin Receptor (GHS-R). UniProt ID: O08725. Species reactivity: Rat. Creossreactivity: Predicted species reactivity based on epitope sequence homology: human, mouse, marmoset, rhesus macaque, platypus, horse. Applications: ELISA, IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide. Handling: Keep on ice at all times.