Learn More
HCL Labels Pre-Labeled GHS Spray Bottles - Custom (Send in your SDS) - 32oz
SDP
Catalog No. NC1974418
Encompass_Preferred
GHS Compliant - Pre-labeled Spray Bottles - 32 oz - These bottles include a pre-affixed GHS hazard communication label, and are ready to be filled (The bottles are empty)Made from chemical resistant HDPE material including a reinforced sprayer - they will not fail, or leak - even after heavy use.Send in your SDS for your custom chemical.Bottles include caps for when a sprayer isn't needed.Bottles have a fully adjustable spray nozzle - 1.4ml per strokeThere is no longer a need to guess dilutions from concentrated liquids. Our 32oz bottles have an easy-to-read 10:1 to 1:1 dilution scale.Labels are made of a durable Adhesive Vinyl that is laminated for chemical and solvent resistance.
Catalog No. NC1974418 Supplier HCL Labels Supplier No. GHSBOTXX32
GHS Compliant - Pre-labeled Spray Bottles - 32 oz - These bottles include a pre-affixed GHS hazard communication label, and are ready to be filled (The bottles are empty)Made from chemical resistant HDPE material including a reinforced sprayer - they will not fail, or leak - even after heavy use.Send in your SDS for your custom chemical.Bottles include caps for when a sprayer isn't needed.Bottles have a fully adjustable spray nozzle - 1.4ml per strokeThere is no longer a need to guess dilutions from concentrated liquids. Our 32oz bottles have an easy-to-read 10:1 to 1:1 dilution scale.Labels are made of a durable Adhesive Vinyl that is laminated for chemical and solvent resistance.