Learn More
Eurofins Discoverx PathHunter GHSR Bioassay Kit, 10-Plate. (93-0242Y3-00142)
Catalog No. 502000060
Encompass
The PathHunter GHSR Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure long-term assay reproducibility, and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-0060 Supplier Eurofins Discoverx Supplier No. 930242Y300142
Add to Cart
Add to Cart
Request a Quote
The PathHunter GHSR Bioassay kit provides a robust, highly sensitive, and easy-to-use functional cell based assay to measure drug potency and detect neutralizing antibodies. The bioassay kit contains all the materials needed for a complete assay including cryopreserved cells, detection reagents, cell plating reagent, positive control agonist, and assay plates. The ready-to-use cryopreserved cells have been manufactured to ensure long-term assay reproducibility, and faster implementation from characterization to lot release. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.