Learn More
Electron Microscopy Sciences Precision Light Guide GI-20-6 with Stay-Put Gooseneck, 20" OAL, 6 mm glass diameter
SDP
Catalog No. 5033272
Encompass_Preferred
Designed for use with microscopes, boroscopes, and endoscopes. Flexible and versatile, the light guides are ideal for a host of scientific, quality control and electronic applications. All of our light guides are backed by a full one-year warranty. Applications in stereochemistry, high/low contrast, optical comparators, IC/Semicondoctor etching and printing, automatic test equipment, profile projectors, and quality control.
Features fiber bundles that have been designed to provide brilliant specimen cross lightning from above and below. Only the finest quality optical class used. End fitting are polished stainless steel.
OAL: 20"
Diameter: 6 mm glass
Catalog No. 50-332-72 Supplier Electron Microscopy Sciences Supplier No. 6795102
Designed for use with microscopes, boroscopes, and endoscopes. Flexible and versatile, the light guides are ideal for a host of scientific, quality control and electronic applications. All of our light guides are backed by a full one-year warranty. Applications in stereochemistry, high/low contrast, optical comparators, IC/Semicondoctor etching and printing, automatic test equipment, profile projectors, and quality control.
Features fiber bundles that have been designed to provide brilliant specimen cross lightning from above and below. Only the finest quality optical class used. End fitting are polished stainless steel.
OAL: 20"
Diameter: 6 mm glass