Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab GI Fix Specialty Prefills
SDP

Catalog No. 23640019
Change view
Click to view available options
Quantity:
40 mL
60 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-640-019 40 mL
23-640-015 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 23-640-019 Supplier StatLab Supplier No. GIFIX0115
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Designed for use on gastrointestinal and liver tissues

  • Produces high quality results on tissues containing large amounts of mucin, inflammatory cells, and cytoplasmic RNA while eliminating the use of hazardous chemicals
  • Available in 20, 40, or 60 mL StatClick prefills in histopaks
  • All Prefill Containers are filled 50 with fixative

Specifications

For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 40 mL
Solution Type Formaldehyde
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.