Learn More
Microbiologics GI Hydration Fluid
SDP
Catalog No. 21402645 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
8 x 1 mL
Catalog No. Quantity
21-402-645 8 x 1 mL
1 options
Catalog No. 21-402-645 Supplier Microbiologics Supplier No. HE0082HF
Add to Cart
Add to Cart
Helix elite™ inactivated standard a buffered saline solution formulated to simulate the composition of common gastrointestinal sample transport media
  • GI Hydration Fluid contains 8 (1.0 mL) vials of hydration fluid.
  • The fluid is a buffered saline solution containing sodium chloride, calcium chloride, and phosphate buffer at relevant concentrations and pH to mimic the composition of common sample transport media for GI testing.
Specifications
Manufacturing Origin United States
Product Type Buffer Solution
Sterility Sterile
Quantity 8 x 1 mL