Learn More
Aqua Solutions GI/NTA Mobile Phase @ 3.15 pH
SDP
Catalog No. NC1802484
Encompass_Preferred
GI/NTA Mobile Phase @ 3.15 pH | 0.03% Sulfuric Acid (7664-93-9) 0.0125% Sodium Hydroxide (1310-73-2) 0.0094% Copper Sulfate Anhydrous (7758-98-7) 99.948% Water
Catalog No. NC1802484 Supplier Aqua Solutions Supplier No. MLU043
GI/NTA Mobile Phase @ 3.15 pH | 0.03% Sulfuric Acid (7664-93-9) 0.0125% Sodium Hydroxide (1310-73-2) 0.0094% Copper Sulfate Anhydrous (7758-98-7) 99.948% Water