Learn More
Maine Molecular Quality Controls Inc FilmArray GI Control Panel M238
SDP
Catalog No. NC0702906
Encompass
The FilmArray GI Control Panel M238 is intended for in vitro use as a quality control to monitor the detection and identification of multiple gastrointestinal pathogens performed with the FilmArray Gastrointestinal (GI) Panel assay. Detection of the viruses, bacteria and protozoa is an important aid to the diagnosis of gastrointestinal illness. The FilmArray GI Control Panel M238 is comprised of 12 tubes, 200µL each, containing synthetic RNA suspended in a non-infectious solution of buffers, preservatives and stabilizers. FilmArray GI Control Panel M238 is composed of 2 controls, FilmArray GI Control M239 and FilmArray GI Control M240, both containing synthetic RNA corresponding to genome segments of pathogens listed in Tables 1 and 2. The synthetic RNA is specifically designed for and intended to be used solely with the BIOFIRE GI Panel and BIOFIRE GI Panel Mid on the BIOFIRE FILMARRAY System.
Catalog No. NC0702906 Supplier Maine Molecular Quality Controls Inc Supplier No. M238
The FilmArray GI Control Panel M238 is intended for in vitro use as a quality control to monitor the detection and identification of multiple gastrointestinal pathogens performed with the FilmArray Gastrointestinal (GI) Panel assay. Detection of the viruses, bacteria and protozoa is an important aid to the diagnosis of gastrointestinal illness. The FilmArray GI Control Panel M238 is comprised of 12 tubes, 200µL each, containing synthetic RNA suspended in a non-infectious solution of buffers, preservatives and stabilizers. FilmArray GI Control Panel M238 is composed of 2 controls, FilmArray GI Control M239 and FilmArray GI Control M240, both containing synthetic RNA corresponding to genome segments of pathogens listed in Tables 1 and 2. The synthetic RNA is specifically designed for and intended to be used solely with the BIOFIRE GI Panel and BIOFIRE GI Panel Mid on the BIOFIRE FILMARRAY System.