Learn More
Medchemexpress LLC GI254023X | 260264-93-5 | MFCD19381832 | 99.6% | 391.50 | C21H33N3O4 | 10 MG
SDP
Catalog No. 5000297194
Encompass_Preferred
GI254023X is a selective small-molecule inhibitor of the metalloproteases ADAM10 and MMP9. It exhibits low-nanomolar potency and is supplied for research use to investigate proteolytic processing, metalloprotease-dependent signaling, and related biological pathways.
  • Low-nanomolar potency against MMP9 and ADAM10 (IC50 2.5 nM and 5.3 nM).
  • High chemical purity (99.6%).
  • Well-characterized molecular properties (C21H33N3O4; MW 391.50).
  • Supplied as a 10 MG research pack suitable for assay development and mechanistic studies.
  • Intended for research use only; not for human or therapeutic use.
Catalog No. 50-002-97194 Supplier Medchemexpress LLC Supplier No. HY1995610MG
GI254023X is a selective small-molecule inhibitor of the metalloproteases ADAM10 and MMP9. It exhibits low-nanomolar potency and is supplied for research use to investigate proteolytic processing, metalloprotease-dependent signaling, and related biological pathways.
  • Low-nanomolar potency against MMP9 and ADAM10 (IC50 2.5 nM and 5.3 nM).
  • High chemical purity (99.6%).
  • Well-characterized molecular properties (C21H33N3O4; MW 391.50).
  • Supplied as a 10 MG research pack suitable for assay development and mechanistic studies.
  • Intended for research use only; not for human or therapeutic use.