Learn More
Selleck Chemical LLC GI254023X 2mg 260264-93-5 GI 4023
SDP
Supplier:  Selleck Chemical LLC S86602MG
Encompass_Preferred
GI254023X (GI 4023) is a selective inhibitor of ADAM10 with 100-fold selectivity for ADAM10 over ADAM17. The IC50 values for recombinant ADAM10 amd ADAM17 are 5.3 nM and 541 nM, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8167