Learn More
ACROBiosystems Varicella zoster virus (strain Oka vaccine) gI&gE Protein, His Tag&Twin-Strep Tag (HPLC-verified)
Catalog No. 502719589
Encompass_Preferred
Varicella-zoster virus (VZV) glycoprotein gI is a type 1 transmembrane glycoprotein which is one component of the heterodimeric gE:gI Fc receptor complex. Like VZV gE, VZV gI was phosphorylated in both VZV-infected cells and gI-transfected cells.
Catalog No. 50-271-9589 Supplier ACROBiosystems Supplier No. GIEV528350UG
Varicella-zoster virus (VZV) glycoprotein gI is a type 1 transmembrane glycoprotein which is one component of the heterodimeric gE:gI Fc receptor complex. Like VZV gE, VZV gI was phosphorylated in both VZV-infected cells and gI-transfected cells.