Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Giant Magnetic Solar System
SCI_ED

Catalog No. S02200 Shop All Learning Resources, Inc Products
Change view
Click to view available options
Grade:
K
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02200 K
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02200 Supplier Learning Resources, Inc Supplier No. LER6040
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Eight planets, Pluto (dwarf planet), sun, Earth's moon, asteroid belt and Activity Guide

Capture students' attention as you model space science concepts on your whiteboard.

  • Full-color, realistically detailed magnets
  • Sun is the largest object

Order Info

Grades K+. Sun measures 10 in. dia. (25.4cm).

Specifications

Grade Level Elementary
Grade K
For Use With (Application) Model space science concepts
Includes Eight planets, Pluto (dwarf planet), sun, Earth's moon, asteroid belt and activity guide
Diameter (English) Outer 10 in. Sun (Largest Object)
Description Giant Magnetic Solar System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.