Learn More
Uline Giant Plastic Stackable Bins - 17 12 x 10 78 x 12 12"", Clear, 4/pk
Catalog No. NC1587528
Encompass
Pack and stack. Holds large, heavy and bulky products, parts and tools. Wide cut-out front opening for easy picking. Large easy-to-grip handles. Stable stacking up to 6 high on floor or shelf. See-through design allows you to easily identify contents.
Catalog No. NC1587528 Supplier Uline Supplier No. S17887PK
Pack and stack. Holds large, heavy and bulky products, parts and tools. Wide cut-out front opening for easy picking. Large easy-to-grip handles. Stable stacking up to 6 high on floor or shelf. See-through design allows you to easily identify contents.